Learn More
Methoxychlor, Thermo Scientific Chemicals
Molecule with estrogenic activity
£34.70 - £127.00
Chemical Identifiers
CAS | 72-43-5 |
---|---|
Molecular Formula | C16H15Cl3O2 |
Molecular Weight (g/mol) | 345.644 |
MDL Number | MFCD00000803 |
InChI Key | IAKOZHOLGAGEJT-UHFFFAOYSA-N |
Synonym | methoxychlor, p,p'-methoxychlor, dmdt, dimethoxy-ddt, methoxy-ddt, methoxcide, metox, maralate, marlate, moxie |
PubChem CID | 4115 |
ChEBI | CHEBI:6842 |
IUPAC Name | 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene |
SMILES | COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455999
|
Thermo Scientific Alfa Aesar
J63839.06 |
5 g |
£34.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465999
|
Thermo Scientific Alfa Aesar
J63839.14 |
25 g |
£127.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molecule with estrogenic activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72-43-5 | |
345.644 | |
IAKOZHOLGAGEJT-UHFFFAOYSA-N | |
4115 | |
1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene |
C16H15Cl3O2 | |
MFCD00000803 | |
methoxychlor, p,p'-methoxychlor, dmdt, dimethoxy-ddt, methoxy-ddt, methoxcide, metox, maralate, marlate, moxie | |
CHEBI:6842 | |
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl |
Specifications
72-43-5 | |
White | |
MFCD00000803 | |
2057367 | |
methoxychlor, p,p'-methoxychlor, dmdt, dimethoxy-ddt, methoxy-ddt, methoxcide, metox, maralate, marlate, moxie | |
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl | |
345.644 | |
CHEBI:6842 | |
Crystalline Powder |
87°C | |
C16H15Cl3O2 | |
UN3077 | |
14,5990 | |
IAKOZHOLGAGEJT-UHFFFAOYSA-N | |
1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene | |
4115 | |
345.65 | |
Methoxychlor |
Safety and Handling
GHS H Statement
H351-H302-H312-H332
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H351
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 200-779-9
missing translation for 'rtecsNumber' : KJ3675000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only