missing translation for 'onlineSavingsMsg'
Learn More

Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 10606802
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH5NO·HCl
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 148°C to 153°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3ONH2·HCl
Quantity 25 g
Beilstein 01, 288
Fieser 01,670
Merck Index 15, 6060
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 83.52
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Methoxylamine hydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methoxylamine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.