missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-α-Methoxyphenylacetic acid, for gravimetric determination of Na, ≥99.0% (T), Honeywell Fluka™
Click to view available options
1 g
Description
Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 69°C to 71°C |
Color | White |
Boiling Point | 165°C (24 hPa) |
Quantity | 1 g |
Molecular Weight (g/mol) | 166.17 g/mol |
Percent Purity | ≥ 99% (T) |
Grade | General Gravimetric Reagents |
Physical Form | Crystalline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'