missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxypolyethylene glycol amine, M.W. 10,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is used as a reagent for pegylation of biomolecules, improvement of solubility and other pharmacological parameters, for the preparation of various conjugates containing polyethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Granular Solid |
Solubility Information | Soluble in water. |
Quantity | 250 mg |
Chemical Name or Material | Methoxypolyethylene glycol amine, M.W. 13,000 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'