missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1-aminocyclopropanecarboxylate hydrochloride, 98%, Thermo Scientific™
£186.12 - £615.23
Chemical Identifiers
CAS | 72784-42-0 |
---|---|
Molecular Formula | C5H9NO2·ClH |
Molecular Weight (g/mol) | 151.59 |
InChI Key | CHSDCWKLSHPKFY-UHFFFAOYSA-N |
Synonym | methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, methyl 1-aminocyclopropane-1-carboxylate hcl, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropanecarboxylate hcl, methyl-1-aminocyclopropanecarboxylate hydrochloride |
PubChem CID | 12760695 |
IUPAC Name | methyl 1-aminocyclopropane-1-carboxylate;hydrochloride |
SMILES | COC(=O)C1(CC1)N.Cl |
Description
Chemical Identifiers
72784-42-0 | |
151.59 | |
methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, methyl 1-aminocyclopropane-1-carboxylate hcl, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropanecarboxylate hcl, methyl-1-aminocyclopropanecarboxylate hydrochloride | |
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride |
C5H9NO2·ClH | |
CHSDCWKLSHPKFY-UHFFFAOYSA-N | |
12760695 | |
COC(=O)C1(CC1)N.Cl |
Specifications
72784-42-0 | |
White | |
Authentic | |
C5H9NO2·ClH | |
CHSDCWKLSHPKFY-UHFFFAOYSA-N | |
methyl 1-aminocyclopropane-1-carboxylate;hydrochloride | |
12760695 | |
98% | |
Methyl 1-aminocyclopropanecarboxylate hydrochloride |
176.4°C to 178.7°C | |
38.37 to 40.33% C, 8.95 to 9.40% | |
98% | |
methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-aminocyclopropane-1-carboxylic acid methyl ester hydrochloride, methyl 1-aminocyclopropane-1-carboxylate hydrochloride, 1,1-accp ome, 1-amino-cyclopropanecarboxylic acid methyl ester hcl, methyl 1-aminocyclopropane-1-carboxylate hcl, cyclopropanecarboxylic acid, 1-amino-, methyl ester, hydrochloride, ksc498c1l, methyl 1-aminocyclopropanecarboxylate hcl, methyl-1-aminocyclopropanecarboxylate hydrochloride | |
COC(=O)C1(CC1)N.Cl | |
151.59 | |
151.59 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Methyl 1-aminocyclopropanecarboxylate hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only