missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Specifications
Melting Point | 172°C |
Color | Pink to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 25 g |
Formula Weight | 283.76 |
Percent Purity | 97% |
Physical Form | Crystalline Powder |
Chemical Name or Material | Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'