missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-cyclohexene-1-carboxylate, 97%

Product Code. 11457246
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11457246

Brand: Thermo Scientific Alfa Aesar B25568.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18448-47-0 | C8H12O2 | 140.182 g/mol

Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases. 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18448-47-0
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.182
MDL Number MFCD00001544
InChI Key KXPWRCPEMHIZGU-UHFFFAOYSA-N
Synonym methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene
PubChem CID 87647
IUPAC Name methyl cyclohexene-1-carboxylate
SMILES COC(=O)C1=CCCCC1

Specifications

Density 1.03
Boiling Point 190°C to 192°C
Flash Point 73°C (165°F)
Refractive Index 1.477
Quantity 5 g
Solubility Information Immiscible with water.
Formula Weight 140.18
Percent Purity 97%
Chemical Name or Material Methyl 1-cyclohexene-1-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.