missing translation for 'onlineSavingsMsg'
Learn More
Methyl 1-hydroxy-1-cyclopropane carboxylate, 90%, Thermo Scientific Chemicals Product Code.: 10671642

Methyl 1-hydroxy-1-cyclopropane carboxylate, 90%, Thermo Scientific Chemicals

Product Code. 10671642
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10671642

Brand: Thermo Scientific Acros 302110050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33689-29-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD02093884
InChI Key KPJWVJURYXOHOO-UHFFFAOYSA-N
Synonym methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl esterShow MoreShow Less
PubChem CID 2733178
IUPAC Name methyl 1-hydroxycyclopropane-1-carboxylate
SMILES COC(=O)C1(O)CC1

Specifications

Color Yellow
Boiling Point 70°C (13.0 mmHg)
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.452
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in most organic solvents
Formula Weight 116
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Methyl 1-hydroxy-1-cyclopropane carboxylate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0368420 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number302110050,302110010
Lot NumberA0359522 Certificate TypeCertificate of Analysis Date10/11/2022 Catalog Number302110050,302110010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 1-hydroxy-1-cyclopropane carboxylate, 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.