Learn More
Methyl 1-hydroxy-1-cyclopropane carboxylate, 90%, Thermo Scientific Chemicals
£109.00 - £343.00
Chemical Identifiers
CAS | 33689-29-1 |
---|---|
Molecular Formula | C5H8O3 |
Molecular Weight (g/mol) | 116.12 |
MDL Number | MFCD02093884 |
InChI Key | KPJWVJURYXOHOO-UHFFFAOYSA-N |
Synonym | methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl ester |
PubChem CID | 2733178 |
IUPAC Name | methyl 1-hydroxycyclopropane-1-carboxylate |
SMILES | COC(=O)C1(O)CC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071831
|
Thermo Scientific Acros
302110010 |
1 g | Glass bottle |
£109.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10671642
|
Thermo Scientific Acros
302110050 |
5 g | Glass bottle |
£343.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33689-29-1 | |
116.12 | |
KPJWVJURYXOHOO-UHFFFAOYSA-N | |
2733178 | |
COC(=O)C1(O)CC1 |
C5H8O3 | |
MFCD02093884 | |
methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl ester | |
methyl 1-hydroxycyclopropane-1-carboxylate |
Specifications
33689-29-1 | |
70°C (13.0 mmHg) | |
Authentic | |
C5H8O3 | |
MFCD02093884 | |
Solubility in water: .. Other solubilities: soluble in most organic solvents | |
COC(=O)C1(O)CC1 | |
116.12 | |
116 | |
Liquid |
Yellow | |
17°C | |
88% min. (GC) | |
1.4470 to 1.452 | |
methyl 1-hydroxycyclopropanecarboxylate, methyl 1-hydroxy-1-cyclopropane carboxylate, methyl 1-hydroxy-1-cyclopropanecarboxylate, 1-hydroxycyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropanecarboxylic acid methyl ester, methyl-1-hydroxy-1-cyclopropane carboxylate, hydroxy-cyclopropanecarboxylic acid methyl ester, 1-hydroxy-cyclopropane-1-carboxylic acid methyl ester, cyclopropanecarboxylic acid, 1-hydroxy-, methyl ester | |
KPJWVJURYXOHOO-UHFFFAOYSA-N | |
methyl 1-hydroxycyclopropane-1-carboxylate | |
2733178 | |
90% | |
Methyl 1-hydroxy-1-cyclopropane carboxylate |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only