missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methylpyrrole-2-carboxylate, 99%, Thermo Scientific Chemicals

Product Code. 11464424
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-methylpyrrole-2-carboxylate is used as chemical and organic Intermediates.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37619-24-2
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00052747
InChI Key APHVGKYWHWFAQV-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-pyrrole-2-carboxylate, methyl1-methyl-1h-pyrrole-2-carboxylate, 1-methyl-1h-pyrrole-2-carboxylic acid methyl ester, methyl 1-methyl-2-pyrrolecarboxylate, 1-methylpyrrole-2-carboxylic acid methyl ester, pubchem12438, acmc-209iue, methylmethylpyrrolecarboxylate, 2-methoxycarbonyl-1-methylpyrrole, methyl 1-methyl-pyrrole-2-carboxylate
PubChem CID 142178
IUPAC Name methyl 1-methylpyrrole-2-carboxylate
SMILES CN1C=CC=C1C(=O)OC

Specifications

Boiling Point 96°C to 98°C (15 mmHg)
Refractive Index 1.522
Quantity 5 g
Beilstein 116477
Solubility Information Slightly soluble in water.
Formula Weight 139.16
Percent Purity 99%
Chemical Name or Material Methyl 1-methylpyrrole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.