missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-difluoro-4-nitrobenzoate, 95%, Thermo Scientific Chemicals

Product Code. 15430189
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924868-81-5
Molecular Formula C8H5F2NO4
Molecular Weight (g/mol) 217.13
MDL Number MFCD08689670
InChI Key XBUVRWIIEREYFL-UHFFFAOYSA-N
Synonym methyl 2,5-difluoro-4-nitrobenzenecarboxylate, methyl2,5-difluoro-4-nitrobenzoate, benzoic acid, 2,5-difluoro-4-nitro-, methyl ester, methyldifluoronitrobenzenecarboxylate, methyl-2,5-difluoro-4-nitrobenzoate, 2,5-difluoro-4-methoxycarbonyl nitrobenzene, 2,5-difluoro-4-nitro-benzoic acid methyl ester
PubChem CID 18526058
IUPAC Name methyl 2,5-difluoro-4-nitrobenzoate
SMILES COC(=O)C1=CC(F)=C(C=C1F)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 217.13
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 2,5-difluoro-4-nitrobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'