missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-amino-4-chlorobenzoate, 95%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Specifications
Melting Point | 65°C to 71°C |
Color | Brown to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Molecular Formula | C8H8ClNO2 |
Quantity | 25 g |
Molecular Weight (g/mol) | 185.607 |
Formula Weight | 185.61 |
Percent Purity | ≥97.5% |
Show More |
Safety and Handling
Product Identifier
- Methyl 2-amino-4-chlorobenzoate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'