Learn More
Methyl 2-amino-4-chlorobenzoate, 95%, Thermo Scientific™
£67.30 - £250.00
Chemical Identifiers
CAS | 5900-58-3 |
---|---|
InChI Key | YPSSCICDVDOEAI-UHFFFAOYSA-N |
Synonym | methyl 4-chloroanthranilate, 2-amino-4-chloro-benzoic acid methyl ester, methyl2-amino-4-chlorobenzoate, benzoic acid, 2-amino-4-chloro-, methyl ester, 2-amino-4-chlorobenzoic acid methyl ester, pubchem4643, methyl 4-chloro-anthranilate, methyl 2-amino4-chlorobenzoate, methyl-2-amino-4-chlorobenzoate |
PubChem CID | 80001 |
IUPAC Name | methyl 2-amino-4-chlorobenzoate |
SMILES | COC(=O)C1=C(C=C(C=C1)Cl)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11965361
|
Thermo Scientific Acros
439230250 |
25 g | Glass bottle | ||||||
11975361
|
Thermo Scientific Acros
439231000 |
100 g | Glass bottle |
£250.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
5900-58-3 | |
methyl 4-chloroanthranilate, 2-amino-4-chloro-benzoic acid methyl ester, methyl2-amino-4-chlorobenzoate, benzoic acid, 2-amino-4-chloro-, methyl ester, 2-amino-4-chlorobenzoic acid methyl ester, pubchem4643, methyl 4-chloro-anthranilate, methyl 2-amino4-chlorobenzoate, methyl-2-amino-4-chlorobenzoate | |
methyl 2-amino-4-chlorobenzoate |
YPSSCICDVDOEAI-UHFFFAOYSA-N | |
80001 | |
COC(=O)C1=C(C=C(C=C1)Cl)N |
Specifications
5900-58-3 | |
Brown to White | |
98% | |
methyl 4-chloroanthranilate, 2-amino-4-chloro-benzoic acid methyl ester, methyl2-amino-4-chlorobenzoate, benzoic acid, 2-amino-4-chloro-, methyl ester, 2-amino-4-chlorobenzoic acid methyl ester, pubchem4643, methyl 4-chloro-anthranilate, methyl 2-amino4-chlorobenzoate, methyl-2-amino-4-chlorobenzoate | |
COC(=O)C1=C(C=C(C=C1)Cl)N | |
185.607 | |
185.61 | |
Crystalline Powder or Powder |
65°C to 71°C | |
Authentic | |
C8H8ClNO2 | |
YPSSCICDVDOEAI-UHFFFAOYSA-N | |
methyl 2-amino-4-chlorobenzoate | |
80001 | |
≥97.5% | |
Methyl 2-amino-4-chlorobenzoate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Methyl 2-amino-4-chlorobenzoate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only