missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate, 97%, Thermo Scientific™

Product Code. 15468475
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 350988-88-4
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.31
MDL Number MFCD01922145
InChI Key AIHYNLWSPTZXGT-UHFFFAOYSA-N
Synonym 2-amino-5-methyl-4-phenyl-thiophene-3-carboxylic acid methyl ester, methyl 2-amino-5-methyl-4-phenyl-3-thiophenecarboxylate, 2-amino-5-methyl-4-phenylthiophene-3-carboxylic acid methyl ester, cbmicro_030702, 3-thiophenecarboxylicacid, 2-amino-5-methyl-4-phenyl-, methyl ester
PubChem CID 734155
IUPAC Name methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
SMILES COC(=O)C1=C(N)SC(C)=C1C1=CC=CC=C1

Specifications

Melting Point 113°C to 114°C
Quantity 5 g
Formula Weight 247.32
Percent Purity 97%
Chemical Name or Material Methyl 2-amino-5-methyl-4-phenylthiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'