Learn More
Methyl 2-chloropropionate, 97%, Thermo Scientific Chemicals
Description
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.
Solubility
Insoluble in water
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | <-10°C |
Density | 1.14 |
Boiling Point | 132°C to 133°C |
Flash Point | 38°C (100°F) |
Refractive Index | 1.417 |
Quantity | 1000 g |
UN Number | UN2933 |
Beilstein | 1421535 |
Solubility Information | Insoluble in water |
Formula Weight | 122.55 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T21G036 | Certificate of Analysis | 23/11/2021 | A18541.30,A18541.0B |
M12H001 | Certificate of Analysis | 23/11/2021 | A18541.30,A18541.0B |
10000624 | Certificate of Analysis | 23/11/2021 | A18541.30,A18541.0B |
14824 | Certificate of Analysis | 23/11/2021 | A18541.30,A18541.0B |
10005608 | Certificate of Analysis | 23/11/2021 | A18541.30,A18541.0B |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.