missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-cyclopentanonecarboxylate, 96%, Thermo Scientific Chemicals
£17.60 - £148.00
Chemical Identifiers
CAS | 10472-24-9 |
---|---|
Molecular Formula | C7H10O3 |
Molecular Weight (g/mol) | 142.15 |
MDL Number | MFCD00001411 |
InChI Key | PZBBESSUKAHBHD-UHFFFAOYSA-N |
Synonym | methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane |
PubChem CID | 66328 |
IUPAC Name | methyl 2-oxocyclopentane-1-carboxylate |
SMILES | COC(=O)C1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347321
|
Thermo Scientific Acros
342090050 |
5 g | Glass bottle |
£17.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10777772
|
Thermo Scientific Acros
342090250 |
25 g | Glass bottle |
£52.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249593
|
Thermo Scientific Acros
342091000 |
100 g | Glass bottle |
£148.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10472-24-9 | |
142.15 | |
PZBBESSUKAHBHD-UHFFFAOYSA-N | |
66328 | |
COC(=O)C1CCCC1=O |
C7H10O3 | |
MFCD00001411 | |
methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane | |
methyl 2-oxocyclopentane-1-carboxylate |
Specifications
10472-24-9 | |
1.0540g/mL | |
77°C | |
95% min. (GC) | |
1.4550 to 1.457 | |
10, 597 | |
methyl 2-oxocyclopentanecarboxylate, methyl 2-cyclopentanonecarboxylate, 2-carbmethoxycyclopentanone, methyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, methyl ester, 2-methoxy carbonyl cyclopentanone, 2-methoxycarbonylcyclopentanone, 2-oxocyclopentanecarboxylic acid methyl ester, cyclopentanone-2-carboxylic acid methyl ester, mehyl-2-oxocyclopentane | |
PZBBESSUKAHBHD-UHFFFAOYSA-N | |
methyl 2-oxocyclopentane-1-carboxylate | |
66328 | |
96% | |
Methyl 2-cyclopentanonecarboxylate |
Colorless to Yellow | |
102°C to 104°C (11.0 mmHg) | |
Authentic | |
C7H10O3 | |
MFCD00001411 | |
1.054 | |
Solubility in water: >100g/L. Other solubilities: miscible in ethanol | |
COC(=O)C1CCCC1=O | |
142.15 | |
142.15 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 233-962-7
RUO – Research Use Only