missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-formylbenzoate, Thermo Scientific™, 97%, Thermo Scientific™

£42.60 - £520.00

Chemical Identifiers

CAS 4122-56-9
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00204231
InChI Key YRMODRRGEUGHTF-UHFFFAOYSA-N
Synonym 2-carbomethoxybenzaldehyde, methyl o-formylbenzoate, phthalaldehydic acid, methyl ester, benzoic acid, formyl-, methyl ester, methyl 2-formylbenzenecarboxylate, benzoic acid, 2-formyl-, methyl ester, methyl 2-formyl-benzoate, methylformylbenzenecarboxylate, methyl-2-formylbenzoate, formyl-benzoic acid methyl ester
PubChem CID 243003
IUPAC Name methyl 2-formylbenzoate
SMILES COC(=O)C1=CC=CC=C1C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10541323
View Documents
Thermo Scientific Maybridge
TL00698DA
1 g
£42.60
1g
Please to purchase this item. Need a web account? Register with us today!
10672073
View Documents
Thermo Scientific Maybridge
TL00698EA
10 g
£138.00
10g
Please to purchase this item. Need a web account? Register with us today!
10089441
View Documents
Thermo Scientific Maybridge
TL00698EE
50 g
£520.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4122-56-9
164.16
YRMODRRGEUGHTF-UHFFFAOYSA-N
243003
COC(=O)C1=CC=CC=C1C=O
C9H8O3
MFCD00204231
2-carbomethoxybenzaldehyde, methyl o-formylbenzoate, phthalaldehydic acid, methyl ester, benzoic acid, formyl-, methyl ester, methyl 2-formylbenzenecarboxylate, benzoic acid, 2-formyl-, methyl ester, methyl 2-formyl-benzoate, methylformylbenzenecarboxylate, methyl-2-formylbenzoate, formyl-benzoic acid methyl ester
methyl 2-formylbenzoate

Specifications

4122-56-9
97%
C9H8O3
MFCD00204231
YRMODRRGEUGHTF-UHFFFAOYSA-N
methyl 2-formylbenzoate
243003
97%
97.0
Amber Glass Bottle
CH3OC(O)C6H4CHO
2-carbomethoxybenzaldehyde, methyl o-formylbenzoate, phthalaldehydic acid, methyl ester, benzoic acid, formyl-, methyl ester, methyl 2-formylbenzenecarboxylate, benzoic acid, 2-formyl-, methyl ester, methyl 2-formyl-benzoate, methylformylbenzenecarboxylate, methyl-2-formylbenzoate, formyl-benzoic acid methyl ester
COC(=O)C1=CC=CC=C1C=O
164.16
164.16
Methyl 2-formylbenzoate
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le

Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.