missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-iodobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11444634
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzoic Acid, methyl ester; 2-Iodobenzoic Acid is a starting material for the preparation of iodosobenzoate, selectivity and environment management. Trivalent iodine compounds perform mild oxidations, oxidative couplings, special iodinations and acetoxylations. Iodobenzoates are used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Insoluble in water.

Notes
Light sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-97-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00016351
InChI Key BXXLTVBTDZXPTN-UHFFFAOYSA-N
Synonym 2-iodobenzoic acid methyl ester, methyl o-iodobenzoate, methyl-2-iodobenzoate, benzoic acid, 2-iodo-, methyl ester, 2-iodo-benzoic acid methyl ester, methyl2-iodobenzoate, benzoic acid, o-iodo-, methyl ester, methyl 2-iodo-benzoate, pubchem10568, acmc-209mp0
PubChem CID 69132
IUPAC Name methyl 2-iodobenzoate
SMILES COC(=O)C1=CC=CC=C1I

Specifications

Density 1.73
Boiling Point 274°C
Flash Point >110°C (230°F)
Refractive Index 1.604
Quantity 50 g
Beilstein 2206859
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material Methyl 2-iodobenzoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'