missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(methylthio)propionate, 99%, Thermo Scientific™
£33.55 - £108.00
Chemical Identifiers
CAS | 61366-76-5 |
---|---|
Molecular Formula | C5H10O2S |
Molecular Weight (g/mol) | 134.193 |
MDL Number | MFCD02683079 |
InChI Key | QMPSBIWJVUCDKV-UHFFFAOYSA-N |
Synonym | methyl 2-methylthio propionate, methyl 2-methylsulfanyl propanoate, methyl 2-methylthio propanoate, dimethyl sulphoniopropionate, methyl 2-methylsulfanyl propanoate #, 2-methylthio propanoic acid methyl ester, 2-methylthiopropionic acid methyl ester, propanoic acid, 2-methylthio-, methyl ester |
PubChem CID | 548372 |
IUPAC Name | methyl 2-methylsulfanylpropanoate |
SMILES | CC(C(=O)OC)SC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsMethyl 2-(methylthio)propionate is used in the contribution of overall aroma perception of the ripe fruit.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
61366-76-5 | |
134.193 | |
QMPSBIWJVUCDKV-UHFFFAOYSA-N | |
548372 | |
CC(C(=O)OC)SC |
C5H10O2S | |
MFCD02683079 | |
methyl 2-methylthio propionate, methyl 2-methylsulfanyl propanoate, methyl 2-methylthio propanoate, dimethyl sulphoniopropionate, methyl 2-methylsulfanyl propanoate #, 2-methylthio propanoic acid methyl ester, 2-methylthiopropionic acid methyl ester, propanoic acid, 2-methylthio-, methyl ester | |
methyl 2-methylsulfanylpropanoate |
Specifications
61366-76-5 | |
1.462 | |
methyl 2-methylthio propionate, methyl 2-methylsulfanyl propanoate, methyl 2-methylthio propanoate, dimethyl sulphoniopropionate, methyl 2-methylsulfanyl propanoate #, 2-methylthio propanoic acid methyl ester, 2-methylthiopropionic acid methyl ester, propanoic acid, 2-methylthio-, methyl ester | |
QMPSBIWJVUCDKV-UHFFFAOYSA-N | |
methyl 2-methylsulfanylpropanoate | |
548372 | |
99% |
C5H10O2S | |
MFCD02683079 | |
Immiscible with water. | |
CC(C(=O)OC)SC | |
134.193 | |
134.2 | |
Methyl 2-(methylthio)propionate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title