missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-octynoate, 98%, Thermo Scientific Chemicals

Product Code. 11424767
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-12-6
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.209
MDL Number MFCD00009530
InChI Key FRLZQXRXIKQFNS-UHFFFAOYSA-N
Synonym methyl 2-octynoate, folione, methyl heptine carbonate, methyl 2-octynate, methyl 2-octinate, 2-octynoic acid, methyl ester, vert de violette, artificial, methyl hept-1-yne-1-carboxylate, methyl pentylacetylenecarboxylate, unii-0ttp6yt2t3
PubChem CID 8092
IUPAC Name methyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OC

Specifications

Density 0.92
Boiling Point 216°C to 218°C
Flash Point 88°C (192°F)
Refractive Index 1.446
Quantity 25 g
Beilstein 1756887
Formula Weight 154.2
Percent Purity 98%
Chemical Name or Material Methyl 2-octynoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'