Learn More
Methyl 2-phenylpyrimidine-5-carboxylate, 97%, Thermo Scientific™
£49.55 - £194.00
Chemical Identifiers
CAS | 64074-29-9 |
---|---|
Molecular Formula | C12H10N2O2 |
Molecular Weight (g/mol) | 214.224 |
MDL Number | MFCD06797254 |
InChI Key | SOMJCOFUAUMQPG-UHFFFAOYSA-N |
Synonym | 2-phenylpyrimidine-5-carboxylic acid methyl ester, methyl2-phenylpyrimidine-5-carboxylate, 2-phenyl-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylicacid,2-phenyl-,methyl ester, 5-pyrimidinecarboxylicacid, 2-phenyl-, methyl ester, 5-pyrimidinecarboxylic acid, 2-phenyl-, methyl ester |
PubChem CID | 11206661 |
IUPAC Name | methyl 2-phenylpyrimidine-5-carboxylate |
SMILES | COC(=O)C1=CN=C(N=C1)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10440754
|
Thermo Scientific Maybridge
CC66322DA |
1 g | Amber Glass Bottle |
£49.55
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386274
|
Thermo Scientific Maybridge
CC66322DE |
5 g | Amber Glass Bottle |
£194.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
64074-29-9 | |
214.224 | |
SOMJCOFUAUMQPG-UHFFFAOYSA-N | |
11206661 | |
COC(=O)C1=CN=C(N=C1)C2=CC=CC=C2 |
C12H10N2O2 | |
MFCD06797254 | |
2-phenylpyrimidine-5-carboxylic acid methyl ester, methyl2-phenylpyrimidine-5-carboxylate, 2-phenyl-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylicacid,2-phenyl-,methyl ester, 5-pyrimidinecarboxylicacid, 2-phenyl-, methyl ester, 5-pyrimidinecarboxylic acid, 2-phenyl-, methyl ester | |
methyl 2-phenylpyrimidine-5-carboxylate |
Specifications
64074-29-9 | |
100.0 | |
C12H10N2O2 | |
2-phenylpyrimidine-5-carboxylic acid methyl ester, methyl2-phenylpyrimidine-5-carboxylate, 2-phenyl-pyrimidine-5-carboxylic acid methyl ester, 5-pyrimidinecarboxylicacid,2-phenyl-,methyl ester, 5-pyrimidinecarboxylicacid, 2-phenyl-, methyl ester, 5-pyrimidinecarboxylic acid, 2-phenyl-, methyl ester | |
COC(=O)C1=CN=C(N=C1)C2=CC=CC=C2 | |
214.224 | |
214.22 | |
Methyl 2-phenylpyrimidine-5-carboxylate |
96.5 | |
97% | |
MFCD06797254 | |
SOMJCOFUAUMQPG-UHFFFAOYSA-N | |
methyl 2-phenylpyrimidine-5-carboxylate | |
11206661 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning