missing translation for 'onlineSavingsMsg'
Learn More
Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, 98%, Thermo Scientific™ Product Code.: 11476536

Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, 98%, Thermo Scientific™

Product Code. 11476536
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11476536

Brand: Thermo Scientific Alfa Aesar B24793.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200573-11-1
Molecular Formula C13H9F3O5S
Molecular Weight (g/mol) 334.27
MDL Number MFCD04039773
InChI Key BDXDWDCWSCPFEW-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, methyl 2-trifluoromethanesulfonyloxy-1-naphthoate, methyl 2-trifluoromethyl sulfonyl oxy-1-naphthoate, methyl 2-trifluoromethanesulfonyloxy naphthalene-1-carboxylate, acmc-20aki5, methyl 2-trifluoromethylsulfonyloxy naphthalene-1-carboxylate, 1-carbomethoxy-2-trifluoromethylsulfonyloxy naphthalene, 2-trifluoromethylsulfonyloxy-1-naphthoic acid methyl ester, 2-trifluoromethylsulfonyloxy naphthalene-1-carboxylic acid methyl esterShow MoreShow Less
PubChem CID 11034997
IUPAC Name methyl 2-(trifluoromethylsulfonyloxy)naphthalene-1-carboxylate
SMILES COC(=O)C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 41°C to 43°C
Quantity 25 g
Formula Weight 334.27
Percent Purity 98%
Chemical Name or Material Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10140303 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10137555 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10145692 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 2-trifluoromethylsulfonyloxy-1-naphthoate, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.