missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-[3-(dimethylamino)propoxy]benzoate, 97%, Thermo Scientific™

Product Code. 10422504
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 921938-75-2
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.299
MDL Number MFCD09702393
InChI Key WGAZXKVAWOAUOI-UHFFFAOYSA-N
Synonym methyl 3-3-dimethylamino propoxy benzoate
PubChem CID 24229605
IUPAC Name methyl 3-[3-(dimethylamino)propoxy]benzoate
SMILES CN(C)CCCOC1=CC=CC(=C1)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.3
Percent Purity 97%
Chemical Name or Material Methyl 3-[3-(dimethylamino)propoxy]benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'