missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,4-dimethoxybenzoate, 98+%, Thermo Scientific Chemicals

Product Code. 11465597
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2150-38-1
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00008430
InChI Key BIGQPYZPEWAPBG-UHFFFAOYSA-N
Synonym methyl veratrate, benzoic acid, 3,4-dimethoxy-, methyl ester, 3,4-dimethoxybenzoic acid methyl ester, veratric acid, methyl ester, methylveratrate, pubchem13009, acmc-1cfay, veratric acid methyl ester, methyl 3,4 dimethoxybenzoate, methyl 3, 4-dimethoxybenzoate
PubChem CID 16522
ChEBI CHEBI:86906
IUPAC Name methyl 3,4-dimethoxybenzoate
SMILES COC(=O)C1=CC=C(OC)C(OC)=C1

Specifications

Melting Point 57°C to 61°C
Boiling Point 283°C
Quantity 250 g
Beilstein 2215244
Formula Weight 196.2
Percent Purity ≥98%
Chemical Name or Material Methyl 3,4-dimethoxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'