missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3,5-dibromo-2-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11486067
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21702-79-4
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00017534
InChI Key KISQISIKTRRMOX-UHFFFAOYSA-N
PubChem CID 624217
IUPAC Name methyl 3,5-dibromo-2-hydroxybenzoate
SMILES COC(=O)C1=CC(Br)=CC(Br)=C1O

Specifications

Melting Point 150°C to 152°C
Quantity 500 g
Synonym methyl 3,5-dibromosalicylate, benzoic acid, 3,5-dibromo-2-hydroxy-, methyl ester, 3,5-dibromo-2-hydroxybenzoic acid methyl ester, pubchem3976, acmc-1cpm5, rarechem al bf 0030, timtec-bb sbb001273, 3,5-dibromo-2-hydroxymethyl benzoate, benzoic acid,3,5-dibromo-2-hydroxy-, methyl ester
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material Methyl 3,5-dibromo-2-hydroxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'