missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, 98%, Thermo Scientific Chemicals

Product Code. 10661273 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10661273

Brand: Thermo Scientific Acros 368280050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1458-18-0
MDL Number MFCD00010431
InChI Key USYMCUGEGUFUBI-UHFFFAOYSA-N
Synonym methyl 3-amino-5,6-dichloro-2-pyrazinecarboxylate, pyrazinecarboxylic acid, 3-amino-5,6-dichloro-, methyl ester, 3-amino-5,6-dichloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-5,6-dichloro-pyrazine-2-carboxylic acid methyl ester, acmc-1bujh, dsstox_cid_31434, dsstox_rid_97320, dsstox_gsid_57645, methyl 3-amino-5,6-dichloropyrazinoate, methyl 3-amino-5,6-dichloropyrazinecarboxylate
PubChem CID 73828
IUPAC Name methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
SMILES COC(=O)C1=C(N=C(C(=N1)Cl)Cl)N

Specifications

Melting Point 227°C to 230°C
Color Brown to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H5Cl2N3O2
Quantity 5 g
Molecular Weight (g/mol) 222.03
Formula Weight 222.03
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 3-amino-5, 6-dichloro-2-pyrazinecarboxylate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.