missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-amino-5-fluorobenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Methyl 3-amino-5-fluorobenzoate is an amino acid widely used in pharmaceuticals and foods.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
Color | White to Yellow |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 169.16 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | Methyl 3-amino-5-fluorobenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'