missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-amino-6-chloropyrazine-2-carboxylate, 97%, Thermo Scientific™
£51.60 - £632.00
Chemical Identifiers
CAS | 1458-03-3 |
---|---|
Molecular Formula | C6H6ClN3O2 |
Molecular Weight (g/mol) | 187.58 |
MDL Number | MFCD00834965 |
InChI Key | JGAJCAJHSHUPGH-UHFFFAOYSA-N |
Synonym | methyl 2-amino-5-chloropyrazine-3-carboxylate, methyl 3-amino-6-chloro-2-pyrazinecarboxylate, 3-amino-6-chloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-6-chloro-, methyl ester, methyl3-amino-6-chloropyrazine-2-carboxylate, ksc524o0p, methyl 2-amino-5-chloro-3-pyrazinylcarboxylate, methyl 3-amino-6-chloro-pyrazine-2-carboxylate, methyl 3-amino-6-chloropyrazinecarboxylate |
PubChem CID | 824637 |
IUPAC Name | methyl 3-amino-6-chloropyrazine-2-carboxylate |
SMILES | COC(=O)C1=NC(Cl)=CN=C1N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1458-03-3 | |
187.58 | |
JGAJCAJHSHUPGH-UHFFFAOYSA-N | |
824637 | |
COC(=O)C1=NC(Cl)=CN=C1N |
C6H6ClN3O2 | |
MFCD00834965 | |
methyl 2-amino-5-chloropyrazine-3-carboxylate, methyl 3-amino-6-chloro-2-pyrazinecarboxylate, 3-amino-6-chloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-6-chloro-, methyl ester, methyl3-amino-6-chloropyrazine-2-carboxylate, ksc524o0p, methyl 2-amino-5-chloro-3-pyrazinylcarboxylate, methyl 3-amino-6-chloro-pyrazine-2-carboxylate, methyl 3-amino-6-chloropyrazinecarboxylate | |
methyl 3-amino-6-chloropyrazine-2-carboxylate |
Specifications
1458-03-3 | |
MFCD00834965 | |
JGAJCAJHSHUPGH-UHFFFAOYSA-N | |
methyl 3-amino-6-chloropyrazine-2-carboxylate | |
824637 | |
97% | |
Methyl 3-amino-6-chloropyrazine-2-carboxylate |
C6H6ClN3O2 | |
methyl 2-amino-5-chloropyrazine-3-carboxylate, methyl 3-amino-6-chloro-2-pyrazinecarboxylate, 3-amino-6-chloro-2-pyrazinecarboxylic acid methyl ester, 3-amino-6-chloro-pyrazine-2-carboxylic acid methyl ester, pyrazinecarboxylic acid, 3-amino-6-chloro-, methyl ester, methyl3-amino-6-chloropyrazine-2-carboxylate, ksc524o0p, methyl 2-amino-5-chloro-3-pyrazinylcarboxylate, methyl 3-amino-6-chloro-pyrazine-2-carboxylate, methyl 3-amino-6-chloropyrazinecarboxylate | |
COC(=O)C1=NC(Cl)=CN=C1N | |
187.58 | |
187.58 | |
Solid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures