missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%

Product Code. p-4228790 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10882641

Brand: Thermo Scientific 432410010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35212-85-2 | C10H9NO2S | 207.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35212-85-2
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.25
MDL Number MFCD00206744
InChI Key VLHHEYMZLXKSQO-UHFFFAOYSA-N
Synonym methyl 3-aminobenzo b thiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, methyl ester, cbmicro_030710, 2-carbomethoxy-3-aminobenzothiophene, methyl 3-aminobenzothiophene-2-carboxylate, methyl-3-aminobenzothiophene-2-carboxylate, 3-amino-2-methoxycarbonyl-1-benzothiophene, methyl 3-amino-benzo b thiophene-2-carboxylate, methyl 3-azanyl-1-benzothiophene-2-carboxylate, amino-benzo b thiophene-2-carboxylic acid methyl ester
PubChem CID 874720
IUPAC Name methyl 3-amino-1-benzothiophene-2-carboxylate
SMILES COC(=O)C1=C(C2=CC=CC=C2S1)N

Specifications

Melting Point 109°C to 111°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-aminobenzo[b]thiophene-2-carboxylate
compliance-icons
Product Identifier
  • Methyl 3-aminobenzo¢b!thiophene-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.