missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chloro-4-methoxybenzoate, 98%, Thermo Scientific™

Product Code. 11410993
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37908-98-8
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017188
InChI Key PINQDVFQCCFACD-UHFFFAOYSA-N
Synonym methyl 3-chloro-p-anisate, 3-chloro-4-methoxybenzoic acid methyl ester, methyl3-chloro-4-methoxybenzoate, benzoic acid, 3-chloro-4-methoxy-, methyl ester, methyl-3-chlor-anisat, acmc-1aehy, rarechem al bf 0700, methyl-3-chloro-4-methoxybenzoate, 2-chloro-4-methoxycarbonyl anisole, 3-cl-4-ch3o-c6h3-cooch3
PubChem CID 123469
IUPAC Name methyl 3-chloro-4-methoxybenzoate
SMILES COC1=C(C=C(C=C1)C(=O)OC)Cl

Specifications

Melting Point 93°C to 97°C
Quantity 250 g
Beilstein 2365816
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material Methyl 3-chloro-4-methoxybenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'