missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-chloroimidazo[1,2-a]pyridine-6-carboxylate, 95%, Thermo Scientific™

Product Code. 15419035
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 900019-34-3
Molecular Formula C9H7ClN2O2
Molecular Weight (g/mol) 210.62
MDL Number MFCD07021500
InChI Key LKRLZIJHMWDMRI-UHFFFAOYSA-N
Synonym methyl 3-chloroimidazo 1,2-a pyridine-6-carboxylate, methyl 3-chloro-4-hydroimidazo 1,2-a pyridine-6-carboxylate, imidazo 1,2-a pyridine-6-carboxylic acid,3-chloro-,methyl ester
PubChem CID 16413558
IUPAC Name methyl 3-chloroimidazo[1,2-a]pyridine-6-carboxylate
SMILES COC(=O)C1=CN2C(Cl)=CN=C2C=C1

Specifications

Melting Point 119°C to 120°C
Quantity 1 g
Formula Weight 210.62
Percent Purity 95%
Chemical Name or Material Methyl 3-chloroimidazo[1,2-a]pyridine-6-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'