missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hexenoate, 95+%, predominantly trans, Thermo Scientific™

Product Code. 10702294
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2396-78-3
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00672771
InChI Key XEAIHUDTEINXFG-SNAWJCMRSA-N
Synonym 3-hexenoic acid, methyl ester, methyl 3-hexenoate, methyl trans-3-hexenoate, methyl e-hex-3-enoate, honeyflor, 3-hexenoic acid, methyl ester, e, unii-7ip0t834pb, methyl hex-3-enoate, methyl e-3-hexenoate, methyl trans 3 hexenoate
PubChem CID 5362782
IUPAC Name methyl (E)-hex-3-enoate
SMILES CCC=CCC(=O)OC

Specifications

Color Colorless to Yellow
Density 0.9150g/mL
Boiling Point 169°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CHCHCH2COOCH3
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.915
Solubility Information Solubility in water: practically insoluble
Formula Weight 128.17
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material Methyl 3-hexenoate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl 3-hexenoate, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.