missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydrazinothiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10475122
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 75681-13-9
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00052080
InChI Key RWGYQWWIXLVGDJ-UHFFFAOYSA-N
Synonym methyl 3-hydrazinothiophene-2-carboxylate, 2-thiophenecarboxylicacid, 3-hydrazinyl-, methyl ester, 3-hydrazino-2-thiophenecarboxylic acid methyl ester, 3-hydrazinothiophene-2-carboxylic acid methyl ester, 3-hydrazino thiophene-2-carboxylic acid methyl ester
PubChem CID 2777595
IUPAC Name methyl 3-hydrazinylthiophene-2-carboxylate
SMILES COC(=O)C1=C(C=CS1)NN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 172.21
Percent Purity 97%
Chemical Name or Material Methyl 3-hydrazinothiophene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'