missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-hydroxy-4-nitrobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11418236
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 713-52-0
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00016997
InChI Key UEGCRFNWTGYVKX-UHFFFAOYSA-N
Synonym benzoic acid, 3-hydroxy-4-nitro-, methyl ester, 3-hydroxy-4-nitrobenzoic acid methyl ester, 3-hydroxy-4-nitro-benzoic acid methyl ester, methyl 3-hydroxy-4-nitro-benzoate, methyl3-hydroxy-4-nitrobenzoate, pubchem20121, acmc-209oia, ksc379e5n, 5-methoxycarbonyl-2-nitrophenol, methyl3-hydroxy-4-nitronemzoate
PubChem CID 294866
IUPAC Name methyl 3-hydroxy-4-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 89°C to 92°C
Quantity 100 g
Beilstein 2583832
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 3-hydroxy-4-nitrobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'