missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-iodo-1H-indole-6-carboxylate, 97%, Thermo Scientific™

Product Code. 10630875
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 850374-98-0
Molecular Formula C10H8INO2
Molecular Weight (g/mol) 301.083
MDL Number MFCD06797504
InChI Key WUFYFIRKVXAESN-UHFFFAOYSA-N
Synonym methyl3-iodo-1h-indole-6-carboxylate, 1h-indole-6-carboxylicacid, 3-iodo-, methyl ester
PubChem CID 7127830
IUPAC Name methyl 3-iodo-1H-indole-6-carboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C(=CN2)I

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 301.08
Percent Purity 97%
Chemical Name or Material Methyl 3-iodo-1H-indole-6-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'