missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Density | 1.085 |
Boiling Point | 164°C |
Flash Point | 60°C (140°F) |
Refractive Index | 1.463 |
Quantity | 25 g |
UN Number | UN1228 |
Beilstein | 1742952 |
Solubility Information | Soluble in water. |
Formula Weight | 120.17 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'