missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3-methylaminocrotonate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.
Solubility
Soluble in water at (54 g/L) (25°C), Calc.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as an intermediate in the synthesis of an analogue to amlodipine, bearing a short amino polyethylene glycol chain.
Solubility
Soluble in water at (54 g/L) (25°C), Calc.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 60°C to 63°C |
Quantity | 5 g |
Solubility Information | Soluble in water at (54g/L) (25°C),Calc. |
Formula Weight | 129.16 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 3-methylaminocrotonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'