missing translation for 'onlineSavingsMsg'
Learn More
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%, Thermo Scientific Chemicals Product Code.: 15498936

Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15498936
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15498936

Brand: Thermo Scientific Alfa Aesar H64467.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188614-01-9
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.25
MDL Number MFCD00449104
InChI Key UFSVRHNRNVVBJR-UHFFFAOYSA-N
PubChem CID 699501
IUPAC Name methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
SMILES COC(=O)C1=CC2=C(SCC(=O)N2)C=C1

Specifications

Melting Point 97°C to 99°C
Quantity 5 g
Beilstein 14311523
Synonym methyl 3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate, methyl-3-oxo-3,4-dihydro-2h-1,4-benzothiazine-6-carboxylate, methyl 3-oxo-3,4-dihydro-2h-benzo b 1,4 thiazine-6-carboxylate, methyl 3-oxo-2h-1,4-benzothiazine-6-carboxylate, methyl 3,4-dihydro-3-oxo-2h-benzo b 1,4 thiazine-6-carboxylate, methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate, pubchem13044, ksc497g3b, methyl-3-oxo-3,4-dihydro-2h-1,, methyl3-oxo-2h-1,4-benzothiazine-6-carboxylate
Formula Weight 223.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.