missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(piperidin-1-ylmethyl)benzoate, 97%, Thermo Scientific™

Product Code. 10361523
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 73278-90-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H19NO2
MDL Number MFCD07772859
Quantity 1 g
Synonym methyl 3-piperidin-1-ylmethyl benzoate, methyl 3-piperidylmethyl benzoate, methyl 3-piperidin-1-yl methyl benzoate, 3-1-piperidinylmethyl benzoic acid, methyl ester, benzoic acid,3-1-piperidinylmethyl-, methyl ester, 2-piperidin-1-yl methyl benzoic acid methyl ester
InChI Key RJOMIJDQGNNOCO-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(=CC=C1)CN2CCCCC2
IUPAC Name methyl 3-(piperidin-1-ylmethyl)benzoate
Molecular Weight (g/mol) 233.311
PubChem CID 7162043
Formula Weight 233.31
Percent Purity 97%
Chemical Name or Material Methyl 3-(piperidin-1-ylmethyl)benzoate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'