missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech., Thermo Scientific Chemicals

£57.89 - £236.18

Chemical Identifiers

CAS 79322-76-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00216476
InChI Key KAXLTAULVFFCNL-UHFFFAOYSA-N
PubChem CID 586417
IUPAC Name methyl 4-(1-hydroxyethyl)benzoate
SMILES CC(C1=CC=C(C=C1)C(=O)OC)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10259470
SDS View Documents
Thermo Scientific Acros
424750010
1 g Glass bottle
£57.89
1g
Please to purchase this item. Need a web account? Register with us today!
10340344
SDS View Documents
Thermo Scientific Acros
424750050
5 g Glass bottle
£236.18
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4-(1-hydroxyethyl)benzoate, 90%, tech., Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.