missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-amino-3-hydroxybenzoate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 118°C to 122°C |
Quantity | 2 g |
Beilstein | 608716 |
Formula Weight | 167.17 |
Percent Purity | 98% |
Chemical Name or Material | Methyl 4-amino-3-hydroxybenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'