missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-aminosalicylate, 97%, Thermo Scientific Chemicals

Product Code. 15422337
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl 4-aminosalicylate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4136-97-4
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00088091
InChI Key QQOXBFUTRLDXDP-UHFFFAOYSA-N
Synonym methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
PubChem CID 77787
ChEBI CHEBI:35089
IUPAC Name methyl 4-amino-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)N)O

Specifications

Quantity 500 g
Solubility Information Slightly soluble in water.
Formula Weight 167.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Methyl 4-aminosalicylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'