missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorosalicylate, 98+%, Thermo Scientific™

Product Code. 11304795
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-04-1
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD02093965
InChI Key WPWUDDDJTIZBGL-UHFFFAOYSA-N
PubChem CID 2775386
IUPAC Name methyl 4-fluoro-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)O

Specifications

Melting Point 38°C to 40°C
Quantity 250 mg
Beilstein 2087920
Synonym methyl 4-fluorosalicylate, 4-fluoro-6-hydroxy-benzoic acid methyl ester, methyl-4-fluoro-2-hydroxybenzoate, benzoic acid, 4-fluoro-2-hydroxy-, methyl ester, 4-fluoro-2-hydroxybenzoic acid methyl ester, pubchem4089, acmc-209zkt, methyl4-fluorosalicylate, ksc493c7h, rarechem al bf 1428
Formula Weight 170.14
Percent Purity ≥98%
Chemical Name or Material Methyl 4-fluorosalicylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'