missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxybenzoate, sodium salt, 99%, Thermo Scientific Chemicals

£165.83 - £449.00

Chemical Identifiers

CAS 5026-62-0
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.13
MDL Number MFCD00016470
InChI Key PESXGULMKCKJCC-UHFFFAOYSA-M
Synonym methylparaben sodium, sodium methylparaben, sodium 4-methoxycarbonyl phenolate, preserval ms, methyl 4-hydroxybenzoate, sodium salt, solparol, bonomold omna, methyl 4-hydroxybenzoate sodium salt, sodium 4-carbomethoxyphenolate, unii-cr6k9c2nhk
PubChem CID 23663626
IUPAC Name sodium;4-methoxycarbonylphenolate
SMILES COC(=O)C1=CC=C(C=C1)[O-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10338370
View Documents Promotion Details
Thermo Scientific Acros
235145000
500 g Plastic bottle
£165.83
500g
Please to purchase this item. Need a web account? Register with us today!
10053673
missing translation for 'viewDocuments'
Thermo Scientific Acros
235140025
2.5 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5026-62-0
174.13
PESXGULMKCKJCC-UHFFFAOYSA-M
23663626
COC(=O)C1=CC=C(C=C1)[O-].[Na+]
C8H7NaO3
MFCD00016470
methylparaben sodium, sodium methylparaben, sodium 4-methoxycarbonyl phenolate, preserval ms, methyl 4-hydroxybenzoate, sodium salt, solparol, bonomold omna, methyl 4-hydroxybenzoate sodium salt, sodium 4-carbomethoxyphenolate, unii-cr6k9c2nhk
sodium;4-methoxycarbonylphenolate

Specifications

5026-62-0
White
Authentic
C8H7NaO3
10,5977
Solubility in water: 333g/L (20°C)
COC(=O)C1=CC=C(C=C1)[O-].[Na+]
174.13
174.13
Powder
>125°C
>100°C
99%
MFCD00016470
methylparaben sodium, sodium methylparaben, sodium 4-methoxycarbonyl phenolate, preserval ms, methyl 4-hydroxybenzoate, sodium salt, solparol, bonomold omna, methyl 4-hydroxybenzoate sodium salt, sodium 4-carbomethoxyphenolate, unii-cr6k9c2nhk
PESXGULMKCKJCC-UHFFFAOYSA-M
sodium;4-methoxycarbonylphenolate
23663626
99%
Methyl 4-hydroxybenzoate, sodium salt, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 225-714-1

missing translation for 'rtecsNumber' : DH2455000

Product Identifier
  • Methyl 4-hydroxybenzoate, sodium salt

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates

RUO – Research Use Only