missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-iodobenzoate, 98%, Thermo Scientific Chemicals

£29.70 - £347.00

Chemical Identifiers

CAS 619-44-3
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
MDL Number MFCD00016353
InChI Key DYUWQWMXZHDZOR-UHFFFAOYSA-N
Synonym methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
PubChem CID 69273
IUPAC Name methyl 4-iodobenzoate
SMILES COC(=O)C1=CC=C(C=C1)I
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10636794
View Documents
Thermo Scientific Acros
414460050
5 g Glass bottle
£29.70
5g
Please to purchase this item. Need a web account? Register with us today!
10085810
View Documents
Thermo Scientific Acros
414460250
25 g Glass bottle
£98.30
25g
Please to purchase this item. Need a web account? Register with us today!
10137420
View Documents
Thermo Scientific Acros
414461000
100 g Glass bottle
£347.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

619-44-3
262.04
DYUWQWMXZHDZOR-UHFFFAOYSA-N
69273
COC(=O)C1=CC=C(C=C1)I
C8H7IO2
MFCD00016353
methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
methyl 4-iodobenzoate

Specifications

619-44-3
Beige to Pink
97.5% min. (GC)
CO2CH3C6H4I
methyl p-iodobenzoate, 4-iodobenzoic acid methyl ester, benzoic acid, 4-iodo-, methyl ester, 4-iodo-benzoic acid methyl ester, benzoic acid, p-iodo-, methyl ester, methyl-4-iodobenzoate, methyl 4-iodo-benzoate, methyl-4-iodo-benzoate, pubchem20378, methyl para-iodobenzoate
COC(=O)C1=CC=C(C=C1)I
262.04
262.04
Crystalline Powder
113°C to 117°C
Authentic
C8H7IO2
MFCD00016353
DYUWQWMXZHDZOR-UHFFFAOYSA-N
methyl 4-iodobenzoate
69273
98%
Methyl 4-iodobenzoate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

Warning

missing translation for 'einecsNumber' : 210-597-1

missing translation for 'tsca' : TSCA

Product Identifier
  • Methyl 4-iodobenzoate

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only