missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
Specifications
Melting Point | 81°C to 86°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min |
Packaging | Glass bottle |
Quantity | 5 g |
Synonym | methyl 4-3-methoxy-3-oxopropanoyl benzoate, methyl 4-methoxycarbonylbenzoylacetate, 4-2-methoxycarbonyl-acetyl-benzoic acid methyl ester, 4-methoxycarbonyl-beta-oxobenzenepropanoic acid methyl ester, benzenepropanoic acid,4-methoxycarbonyl-a-oxo-, methyl ester, acmc-20efie, methyl 4-methoxycarbonyl benzoyl acetate, methyl4-methoxycarbonylbenzoylacetate, 4-methoxycarbonylbenzoyl acetic acid methyl ester, methyl 3-4-methoxycarbonyl phenyl-3-oxopropanoate |
Solubility Information | Solubility in water: insoluble in water. Other solubilities: slightly soluble in acetone,methanol |
Formula Weight | 236.22 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- Methyl 4-methoxycarbonylbenzoylacetate
- Danger
- Acute toxicity Category 3
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'