Learn More
Methyl 4-methoxycarbonylbenzoylacetate, 95%, Thermo Scientific™
£30.90 - £75.40
Chemical Identifiers
CAS | 22027-52-7 |
---|---|
Molecular Formula | C13H14O5 |
Molecular Weight (g/mol) | 250.25 |
MDL Number | MFCD00800637 |
InChI Key | GFDCYWYWNSTMDX-UHFFFAOYSA-N |
PubChem CID | 3864207 |
IUPAC Name | methyl 4-(3-methoxy-3-oxopropanoyl)benzoate |
SMILES | CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10297710
|
Thermo Scientific Acros
325710010 |
1 g | Glass bottle |
£30.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10299603
|
Thermo Scientific Acros
325710050 |
5 g | Glass bottle |
£75.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
22027-52-7 | |
250.25 | |
GFDCYWYWNSTMDX-UHFFFAOYSA-N | |
methyl 4-(3-methoxy-3-oxopropanoyl)benzoate |
C13H14O5 | |
MFCD00800637 | |
3864207 | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC |
Specifications
22027-52-7 | |
White to Yellow | |
94% min | |
MFCD00800637 | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in acetone,methanol | |
CCOC(=O)CC(=O)C1=CC=C(C=C1)C(=O)OC | |
250.25 | |
236.22 | |
Powder |
81°C to 86°C | |
Authentic | |
C13H14O5 | |
methyl 4-3-methoxy-3-oxopropanoyl benzoate, methyl 4-methoxycarbonylbenzoylacetate, 4-2-methoxycarbonyl-acetyl-benzoic acid methyl ester, 4-methoxycarbonyl-beta-oxobenzenepropanoic acid methyl ester, benzenepropanoic acid,4-methoxycarbonyl-a-oxo-, methyl ester, acmc-20efie, methyl 4-methoxycarbonyl benzoyl acetate, methyl4-methoxycarbonylbenzoylacetate, 4-methoxycarbonylbenzoyl acetic acid methyl ester, methyl 3-4-methoxycarbonyl phenyl-3-oxopropanoate | |
GFDCYWYWNSTMDX-UHFFFAOYSA-N | |
methyl 4-(3-methoxy-3-oxopropanoyl)benzoate | |
3864207 | |
95% | |
Methyl 4-methoxycarbonylbenzoylacetate |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Toxic in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- Methyl 4-methoxycarbonylbenzoylacetate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only