missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-methyl-3-nitrobenzoate, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 49°C to 53°C |
Molecular Formula | C9H9NO4 |
Quantity | 25 g |
Synonym | methyl 3-nitro-4-methylbenzoate, 4-methyl-3-nitrobenzoic acid methyl ester, benzoic acid, 4-methyl-3-nitro-, methyl ester, methyl 3-nitro-p-toluate, 4-methyl-3-nitro-benzoic acid methyl ester, pubchem10934, acmc-1bfci, ksc497g4t, methyl4-methyl-3-nitrobenzoate, 4-methoxycarbonyl-2-nitrotoluene |
Solubility Information | Soluble in methanol. |
InChI Key | YFPBHPCMYFCRKS-UHFFFAOYSA-N |
SMILES | CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-] |
IUPAC Name | methyl 4-methyl-3-nitrobenzoate |
Molecular Weight (g/mol) | 195.17 |
PubChem CID | 81816 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'