missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-(methylamino)benzoate, 98%, Thermo Scientific Chemicals
£40.60 - £453.00
Chemical Identifiers
CAS | 18358-63-9 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.192 |
MDL Number | MFCD00017198 |
InChI Key | LLAMGYUWYUMHCH-UHFFFAOYSA-N |
Synonym | methyl 4-methylamino benzoate, methyl 4 methylamino benzoate, benzoic acid, 4-methylamino-, methyl ester, acmc-209eks, methyl 4-methylaninobenzoate, 4-carbomethoxy-n-methylaniline, methyl 4-methylamino-benzoate, methyl-4-methylaminobenzoate, 4-methoxycarbonyl-n-methylaniline, 4-methylamino-benzoic acid methylester |
PubChem CID | 140375 |
IUPAC Name | methyl 4-(methylamino)benzoate |
SMILES | CNC1=CC=C(C=C1)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11435533
|
Thermo Scientific Alfa Aesar
A13460.06 |
5 g |
£40.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11445533
|
Thermo Scientific Alfa Aesar
A13460.14 |
25 g |
£151.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455533
|
Thermo Scientific Alfa Aesar
A13460.22 |
100 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilityIt is slightly soluble in water. (1.2 g/L) (25°C insoluble in hexane.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Chemical Identifiers
18358-63-9 | |
165.192 | |
LLAMGYUWYUMHCH-UHFFFAOYSA-N | |
140375 | |
CNC1=CC=C(C=C1)C(=O)OC |
C9H11NO2 | |
MFCD00017198 | |
methyl 4-methylamino benzoate, methyl 4 methylamino benzoate, benzoic acid, 4-methylamino-, methyl ester, acmc-209eks, methyl 4-methylaninobenzoate, 4-carbomethoxy-n-methylaniline, methyl 4-methylamino-benzoate, methyl-4-methylaminobenzoate, 4-methoxycarbonyl-n-methylaniline, 4-methylamino-benzoic acid methylester | |
methyl 4-(methylamino)benzoate |
Specifications
18358-63-9 | |
C9H11NO2 | |
2803327 | |
It is slightly soluble in water. (1.2g/L) (25°C insoluble in hexane. | |
CNC1=CC=C(C=C1)C(=O)OC | |
165.192 | |
165.19 | |
Methyl 4-(methylamino)benzoate |
92°C to 97°C | |
MFCD00017198 | |
methyl 4-methylamino benzoate, methyl 4 methylamino benzoate, benzoic acid, 4-methylamino-, methyl ester, acmc-209eks, methyl 4-methylaninobenzoate, 4-carbomethoxy-n-methylaniline, methyl 4-methylamino-benzoate, methyl-4-methylaminobenzoate, 4-methoxycarbonyl-n-methylaniline, 4-methylamino-benzoic acid methylester | |
LLAMGYUWYUMHCH-UHFFFAOYSA-N | |
methyl 4-(methylamino)benzoate | |
140375 | |
98% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only