missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-n-butoxybenzoate, 98%, Thermo Scientific™

Product Code. 11418247
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4906-25-6
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.26
MDL Number MFCD00070801
InChI Key PUHCYTHGLBCWEA-UHFFFAOYSA-N
Synonym methyl 4-n-butoxybenzoate, benzoic acid, 4-butoxy-, methyl ester, 4-butoxybenzoic acid methyl ester, methyl 4-n-butyloxybenzoate, rarechem al bf 0089, 4-butoxy-benzoic acid methyl ester, labotest-bb lt00847894, benzoic acid, 4-butyloxy-, methyl ester
PubChem CID 4226083
IUPAC Name methyl 4-butoxybenzoate
SMILES CCCCOC1=CC=C(C=C1)C(=O)OC

Specifications

Boiling Point 188°C to 190°C (30 mmHg)
Refractive Index 1.517
Quantity 50 g
Beilstein 2099767
Formula Weight 208.26
Percent Purity 98%
Chemical Name or Material Methyl 4-n-butoxybenzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'